missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzaldehyde 98.0+%, TCI America™

Catalog No. E032525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E032525ML 25 mL
E0325100ML 100 mL
E0325500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E032525ML Supplier TCI America Supplier No. E032525ML

Chemical Identifiers

CAS 4748-78-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00006956
InChI Key QNGNSVIICDLXHT-UHFFFAOYSA-N
Synonym p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
PubChem CID 20861
IUPAC Name 4-ethylbenzaldehyde
SMILES CCC1=CC=C(C=C1)C=O

Specifications

Color Colorless
Boiling Point 221°C
Quantity 25 mL
Formula Weight 134.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylbenzaldehyde
EINECSNumber (3)-3555
TSCA Yes