missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzaldehyde, 98%

CAS: 4748-78-1 | C9H10O | 134.18 g/mol

$120.49 - $120.49

Chemical Identifiers

CAS 4748-78-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00006956
InChI Key QNGNSVIICDLXHT-UHFFFAOYSA-N
PubChem CID 20861
IUPAC Name 4-ethylbenzaldehyde
SMILES CCC1=CC=C(C=C1)C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC378950250
SDS View Documents
Thermo Scientific Chemicals
378950250
25 g Glass bottle
Each for $120.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4748-78-1
134.18
QNGNSVIICDLXHT-UHFFFAOYSA-N
4-ethylbenzaldehyde
C9H10O
MFCD00006956
20861
CCC1=CC=C(C=C1)C=O

Specifications

4748-78-1
100.0
0.9700g/mL
92°C
97.5% min. (GC)
C9H10O
CH3CH2C6H4CHO
25 g
p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
CCC1=CC=C(C=C1)C=O
134.18
134.18
Liquid
97.5
Colorless to Yellow
221.0°C
Authentic
Glass bottle
1.5370 to 1.5410
MFCD00006956
0.97
QNGNSVIICDLXHT-UHFFFAOYSA-N
4-ethylbenzaldehyde
20861
98%
4-Ethylbenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-268-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only