missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylbenzaldehyde, 97%

Catalog No. AAB2064514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2064514 25 g
AAB2064522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2064514 Supplier Thermo Scientific Chemicals Supplier No. B2064514

CAS: 4748-78-1 | C9H10O | 134.178 g/mol

It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a perfuming agent in cosmetics industry. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 4748-78-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00006956
InChI Key QNGNSVIICDLXHT-UHFFFAOYSA-N
Synonym p-ethylbenzaldehyde, benzaldehyde, 4-ethyl, 4-ethyl benzaldehyde, 4-ethyl-benzaldehyde, ebal, ethyl benzaldehyde, ethyl-benzaldehyde, unii-289ppw3sg4, benzaldehyde,4-ethyl, ethylbenzaldehyde, p
PubChem CID 20861
IUPAC Name 4-ethylbenzaldehyde
SMILES CCC1=CC=C(C=C1)C=O

Specifications

Density 0.979
Boiling Point 220°C to 221°C
Flash Point 92°C (197°F)
Odor Characteristic
Refractive Index 1.539
Quantity 25 g
Beilstein 636027
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 4-Ethylbenzaldehyde
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 225-268-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only