missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylanisole 98.0+%, TCI America™

Catalog No. E043625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E043625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E043625ML Supplier TCI America Supplier No. E043625ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1515-95-3
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00015176
InChI Key HDNRAPAFJLXKBV-UHFFFAOYSA-N
Synonym 4-ethylanisole, p-ethylanisole, benzene, 1-ethyl-4-methoxy, anisole, p-ethyl, p-ethylanisol, unii-i71p5qg20s, 4-ethyl-1-methoxybenzene, 4-ethyl anisole, 4-ethylmethoxybenzene, acmc-1btzv
PubChem CID 73690
IUPAC Name 1-ethyl-4-methoxybenzene
SMILES CCC1=CC=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 195°C
Quantity 25 mL
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethylanisole
EINECSNumber (3)-1035
TSCA No