missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylanisole, 98+%

Catalog No. AAA1039109
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1039109 10 g
AAA1039118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1039109 Supplier Thermo Scientific Chemicals Supplier No. A1039109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1515-95-3 | C9H12O | 136.194 g/mol

It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used the synthesis of 1-Methoxy-4-ethyl-1,4-cyclohexadiene. It is also applied in the ligand-assisted, copper-catalyzed enantioselective benzylic amination.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1515-95-3
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00015176
InChI Key HDNRAPAFJLXKBV-UHFFFAOYSA-N
Synonym 4-ethylanisole, p-ethylanisole, benzene, 1-ethyl-4-methoxy, anisole, p-ethyl, p-ethylanisol, unii-i71p5qg20s, 4-ethyl-1-methoxybenzene, 4-ethyl anisole, 4-ethylmethoxybenzene, acmc-1btzv
PubChem CID 73690
IUPAC Name 1-ethyl-4-methoxybenzene
SMILES CCC1=CC=C(C=C1)OC

Specifications

Density 0.962
Boiling Point 195°C
Flash Point 79°C (174°F)
Refractive Index 1.5085
Quantity 10 g
Beilstein 774365
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material 4-Ethylanisole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only