missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethylaniline 99.0+%, TCI America™
$63.38 - $671.06
Chemical Identifiers
| CAS | 589-16-2 |
|---|---|
| Molecular Formula | C8H11N |
| Molecular Weight (g/mol) | 121.18 |
| MDL Number | MFCD00007921 |
| InChI Key | HRXZRAXKKNUKRF-UHFFFAOYSA-N |
| Synonym | p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060 |
| PubChem CID | 11504 |
| IUPAC Name | 4-ethylaniline |
| SMILES | CCC1=CC=C(N)C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
E006125ML
|
TCI America
E006125ML |
25 mL |
Each for $63.38
|
|
|||||
|
E0061500ML
|
TCI America
E0061500ML |
500 mL |
Each for $671.06
|
|
|||||
Chemical Identifiers
| 589-16-2 | |
| 121.18 | |
| HRXZRAXKKNUKRF-UHFFFAOYSA-N | |
| 11504 | |
| CCC1=CC=C(N)C=C1 |
| C8H11N | |
| MFCD00007921 | |
| p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060 | |
| 4-ethylaniline |
Specifications
| 589-16-2 | |
| Yellow | |
| C8H11N | |
| 25 mL | |
| p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060 | |
| CCC1=CC=C(N)C=C1 | |
| 121.18 | |
| 121.18 | |
| Liquid |
| -5°C | |
| 214°C | |
| MFCD00007921 | |
| 2810 | |
| HRXZRAXKKNUKRF-UHFFFAOYSA-N | |
| 4-ethylaniline | |
| 11504 | |
| ≥99.0% (GC,T) | |
| 4-Ethylaniline |
Safety and Handling
EINECSNumber : (3)-0206
RTECSNumber : BX9900000
TSCA : Yes