accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 589-16-2

IUPAC Name: 4-ethylaniline
Synonyms: P-ethylaniline
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: HRXZRAXKKNUKRF-UHFFFAOYSA-N
SMILES: CCC1=CC=C(N)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)