missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylaniline 99.0+%, TCI America™

Catalog No. E006125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E006125ML 25 mL
E0061500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E006125ML Supplier TCI America Supplier No. E006125ML

Chemical Identifiers

CAS 589-16-2
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007921
InChI Key HRXZRAXKKNUKRF-UHFFFAOYSA-N
Synonym p-ethylaniline, benzenamine, 4-ethyl, 4-ethylphenylamine, aniline, 4-ethyl, p-ethylaminobenzene, 1-amino-4-ethylbenzene, 4-aminoethylbenzene, aniline, p-ethyl, unii-mhx72w1zqv, ccris 5060
PubChem CID 11504
IUPAC Name 4-ethylaniline
SMILES CCC1=CC=C(N)C=C1

Specifications

Melting Point -5°C
Color Yellow
Boiling Point 214°C
Quantity 25 mL
UN Number 2810
Formula Weight 121.18
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Ethylaniline
EINECSNumber (3)-0206
RTECSNumber BX9900000
TSCA Yes