missing translation for 'onlineSavingsMsg'
Learn More

4'-Ethylacetophenone 97.0+%, TCI America™

Catalog No. E0318500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0318500ML 500 mL
E031825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0318500ML Supplier TCI America Supplier No. E0318500ML

Chemical Identifiers

CAS 937-30-4
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00009262
InChI Key NODGRWCMFMEGJH-UHFFFAOYSA-N
Synonym 4'-ethylacetophenone, 1-4-ethylphenyl ethanone, p-ethylacetophenone, 4-ethylacetophenone, ethanone, 1-4-ethylphenyl, p-acetylethylbenzene, acetophenone, 4'-ethyl, p-ethylphenyl methyl ketone, 1-4-ethylphenyl ethan-1-one
PubChem CID 13642
IUPAC Name 1-(4-ethylphenyl)ethanone
SMILES CCC1=CC=C(C=C1)C(=O)C

Specifications

Color Yellow
Boiling Point 239°C
Quantity 500 mL
Formula Weight 148.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-Ethylacetophenone
EINECSNumber (3)-3540
TSCA Yes