accessibility menu, dialog, popup

UserName

H3CCH3O

CAS RN 937-30-4

IUPAC Name: 1-(4-ethylphenyl)ethan-1-one
Synonyms: 4-ethylacetophenone
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: NODGRWCMFMEGJH-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results