missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Ethyl-o-xylene 99.0+%, TCI America™
Supplier: TCI America E028101ML
Specifications
| 4-Ethyl-o-xylene | |
| 934-80-5 | |
| 190°C | |
| Clear Liquid at 20°C | |
| MFCD00059234 | |
| 4-ethyl-o-xylene, 1,2-dimethyl-4-ethylbenzene, 2-methyl-p-ethyltoluene, 3,4-dimethylethylbenzene, o-xylene, 4-ethyl, benzene, 4-ethyl-1,2-dimethyl, acmc-209rm3, 1-ethyl-3,4-dimethylbenzene, 3,4-dimethyl-1-ethylbenzene, 1,2-dimethyl-4-ethyl benzene | |
| CCC1=CC(=C(C=C1)C)C | |
| 134.222 | |
| 134.22 |
| -67°C | |
| Yellow | |
| 0.1 mL | |
| C10H14 | |
| 3295 | |
| SBUYFICWQNHBCM-UHFFFAOYSA-N | |
| 4-ethyl-1,2-dimethylbenzene | |
| 13629 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 934-80-5 | |
| 134.222 | |
| SBUYFICWQNHBCM-UHFFFAOYSA-N | |
| 13629 | |
| CCC1=CC(=C(C=C1)C)C |
| C10H14 | |
| MFCD00059234 | |
| 4-ethyl-o-xylene, 1,2-dimethyl-4-ethylbenzene, 2-methyl-p-ethyltoluene, 3,4-dimethylethylbenzene, o-xylene, 4-ethyl, benzene, 4-ethyl-1,2-dimethyl, acmc-209rm3, 1-ethyl-3,4-dimethylbenzene, 3,4-dimethyl-1-ethylbenzene, 1,2-dimethyl-4-ethyl benzene | |
| 4-ethyl-1,2-dimethylbenzene |
Safety and Handling
EINECSNumber : (3)-3427
TSCA : Yes