missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-4-methylmorpholinium Bromide 97.0+%, TCI America™

Catalog No. p-7129374
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
E11665G 5 g
E116625G 25 g
2 options

Catalog No. E11665G

Supplier: TCI America E11665G

Only null left

Chemical Identifiers

CAS 65756-41-4
Molecular Formula C7H16BrNO
Molecular Weight (g/mol) 210.115
MDL Number MFCD03095385
InChI Key ACQZVWGQFXXTIX-UHFFFAOYSA-M
PubChem CID 103453
IUPAC Name 4-ethyl-4-methylmorpholin-4-ium;bromide
SMILES CC[N+]1(CCOCC1)C.[Br-]

Specifications

Melting Point 177°C
Quantity 5 g
Formula Weight 210.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethyl-4-methylmorpholinium Bromide
EINECSNumber (5)-5971
TSCA Yes