accessibility menu, dialog, popup

UserName

BrCH3NH3CO

CAS RN 65756-41-4

IUPAC Name: 4-ethyl-4-methylmorpholin-4-ium bromide
Synonyms: 4-ethyl-4-methylmorpholin-4-ium bromide
Molecular Weight (g/mol): 210.12
Molecular Formula: C7H16BrNO
InChi Key: ACQZVWGQFXXTIX-UHFFFAOYSA-M
SMILES: [Br-].CC[N+]1(C)CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results