missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-2-methoxyphenol, 98%

Catalog No. p-7021768
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1423906 5 g
AAA1423914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1423906 Supplier Thermo Scientific Chemicals Supplier No. A1423906
Only null left

CAS: 2785-89-9 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2785-89-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00038714
InChI Key CHWNEIVBYREQRF-UHFFFAOYSA-N
Synonym 4-ethylguaiacol, p-ethylguaiacol, 2-methoxy-4-ethylphenol, phenol, 4-ethyl-2-methoxy, homocresol, guaiacyl ethane, 4-ethyl-2-methoxy-phenol, 4-hydroxy-3-methoxy ethylbenzene, unii-c9nfd83bj5, guaiacol, 4-ethyl
PubChem CID 62465
IUPAC Name 4-ethyl-2-methoxyphenol
SMILES CCC1=CC=C(O)C(OC)=C1

Specifications

Melting Point 15°C
Density 1.064
Boiling Point 230°C
Flash Point 107°C (225°F)
Refractive Index 1.528
Quantity 5 g
Beilstein 2045329
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-Ethyl-2-methoxyphenol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-500-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only