missing translation for 'onlineSavingsMsg'
Learn More

4-Ethyl-1-octyn-3-ol 97.0+%, TCI America™

Catalog No. E027025ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E027025ML 25 mL
E0270500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E027025ML Supplier TCI America Supplier No. E027025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5877-42-9
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00015262
InChI Key CUUQUEAUUPYEKK-UHFFFAOYNA-N
Synonym (2-Ethyl-1-hydroxyhexyl)acetylene
PubChem CID 93012
IUPAC Name 4-ethyloct-1-yn-3-ol
SMILES CCCCC(CC)C(O)C#C

Specifications

Color Yellow
Boiling Point 126°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethyl-1-octyn-3-ol