accessibility menu, dialog, popup

UserName

H3CCH3OHCH

CAS RN 5877-42-9

IUPAC Name: 4-ethyloct-1-yn-3-ol
Synonyms: 1-octyn-3-ol, 4-ethyl-
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: CUUQUEAUUPYEKK-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(O)C#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results