missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenylacetic acid, 98%

Catalog No. p-7080561
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1012909 10 g
AAA1012914 25 g
AAA1012918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1012909 Supplier Thermo Scientific Chemicals Supplier No. A1012909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4919-33-9 | C10H12O3 | 180.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4919-33-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00004346
InChI Key ZVVWZNFSMIFGEP-UHFFFAOYSA-N
Synonym 4-ethoxyphenylacetic acid, 2-4-ethoxyphenyl acetic acid, 4-ethoxyphenyl acetic acid, benzeneacetic acid, 4-ethoxy, p-ethoxyphenylacetic acid, para-ethoxyphenylacetic acid, acmc-20amjb, 4-ethoxyphenylaceticacid, enamine_004828, p-ethoxyphenyl acetic acid
PubChem CID 78631
IUPAC Name 2-(4-ethoxyphenyl)acetic acid
SMILES CCOC1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 87°C to 91°C
Quantity 10 g
Beilstein 1103143
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxyphenylacetic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-545-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only