missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Ethoxyphenyl)urea 98.0+%, TCI America™
Supplier: TCI America E11711G
Specifications
| (4-Ethoxyphenyl)urea | |
| 170°C | |
| MFCD00025431 | |
| GGLIEWRLXDLBBF-UHFFFAOYSA-N | |
| (4-ethoxyphenyl)urea | |
| 9013 | |
| 180.21 | |
| Crystalline Powder |
| 150-69-6 | |
| C9H12N2O2 | |
| 1 g | |
| CCOC1=CC=C(NC(N)=O)C=C1 | |
| 180.21 | |
| CHEBI:82462 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 150-69-6 | |
| 180.21 | |
| GGLIEWRLXDLBBF-UHFFFAOYSA-N | |
| CHEBI:82462 | |
| CCOC1=CC=C(NC(N)=O)C=C1 |
| C9H12N2O2 | |
| MFCD00025431 | |
| 9013 | |
| (4-ethoxyphenyl)urea |
Safety and Handling
RTECSNumber : YT2275000
TSCA : No
Recommended Storage : Refrigerator