missing translation for 'onlineSavingsMsg'
Learn More

(4-Ethoxyphenyl)urea 98.0+%, TCI America™

Catalog No. E1171200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E1171200MG 200 mg
E11711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E1171200MG Supplier TCI America Supplier No. E1171200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-69-6
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00025431
InChI Key GGLIEWRLXDLBBF-UHFFFAOYSA-N
PubChem CID 9013
ChEBI CHEBI:82462
IUPAC Name (4-ethoxyphenyl)urea
SMILES CCOC1=CC=C(NC(N)=O)C=C1

Specifications

Melting Point 170°C
Quantity 200 mg
Formula Weight 180.21
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (4-Ethoxyphenyl)urea
RTECSNumber YT2275000
TSCA No
Recommended Storage Refrigerator