missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenyl isothiocyanate, 98%

Catalog No. AAA1890206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1890206 5 g
AAA1890214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1890206 Supplier Thermo Scientific Chemicals Supplier No. A1890206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3460-49-9 | C9H9NOS | 179.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3460-49-9
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00041096
InChI Key REEKOMRZYJXXNR-UHFFFAOYSA-N
Synonym 4-ethoxyphenyl isothiocyanate, 4-isothiocyanatophenetol, p-ethoxyphenyl isothiocyanate, 1-ethoxy-4-isothiocyanato-benzene, benzene, 1-ethoxy-4-isothiocyanato, 4-ethoxyphenylisothiocyanate, 4-ethoxybenzenisothiocyanate, acmc-20amjc, pubchem9560, p-ethoxyphenylisothiocyanate
PubChem CID 77006
IUPAC Name 1-ethoxy-4-isothiocyanatobenzene
SMILES CCOC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 57°C to 61°C
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 637526
Sensitivity Moisture sensitive
Formula Weight 179.24
Percent Purity 98%
Chemical Name or Material 4-Ethoxyphenyl isothiocyanate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 222-410-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only