missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)phenyl isothiocyanate, 97%

Catalog No. p-7045937
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1252204 2 g
AAL1252209 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1252204 Supplier Thermo Scientific Chemicals Supplier No. L1252204
Only null left

CAS: 1205-06-7 | C10H9NO2S | 207.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1205-06-7
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.247
MDL Number MFCD00022058
InChI Key MLOJHUCMCKBDLV-UHFFFAOYSA-N
Synonym 4-ethoxycarbonylphenyl isothiocyanate, 4-ethoxycarbonylphenylisothiocyanate, benzoic acid, 4-isothiocyanato-, ethyl ester, 4-ethoxycarbonyl phenyl isothiocyanate, ethyl p-isothiocyanatobenzoate, 4-ethoxycarbonylphenyl-isothiocyanate, benzoic acid,4-isothiocyanato-, ethyl ester, acmc-1brcn, p-carbethoxyphenylisothiocyanate, #
PubChem CID 71003
IUPAC Name ethyl 4-isothiocyanatobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 52°C to 56°C
Boiling Point 123°C to 127°C (1 mmHg)
Odor Irritating
Quantity 2 g
UN Number UN2923
Beilstein 2213400
Sensitivity Moisture sensitive
Formula Weight 207.25
Percent Purity 97%
Chemical Name or Material 4-(Ethoxycarbonyl)phenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
EINECSNumber 214-880-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only