missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Ethoxybenzylidene-4-butylaniline 99.0+%, TCI America™
$193.86 - $1841.68
Chemical Identifiers
| CAS | 29743-08-6 |
|---|---|
| Molecular Formula | C19H23NO |
| Molecular Weight (g/mol) | 281.399 |
| MDL Number | MFCD00009465 |
| InChI Key | DBOAVDSSZWDGTH-UHFFFAOYSA-N |
| Synonym | EBBA |
| PubChem CID | 94406 |
| IUPAC Name | N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine |
| SMILES | CCCCC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OCC |
Chemical Identifiers
| 29743-08-6 | |
| 281.399 | |
| DBOAVDSSZWDGTH-UHFFFAOYSA-N | |
| 94406 | |
| CCCCC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OCC |
| C19H23NO | |
| MFCD00009465 | |
| EBBA | |
| N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine |
Specifications
| 29743-08-6 | |
| C19H23NO | |
| 25 g | |
| DBOAVDSSZWDGTH-UHFFFAOYSA-N | |
| N-(4-butylphenyl)-1-(4-ethoxyphenyl)methanimine | |
| 94406 | |
| ≥99.0% (T) | |
| 4′-Ethoxybenzylidene-4-butylaniline |
| White-Yellow | |
| MFCD00009465 | |
| EBBA | |
| CCCCC1=CC=C(C=C1)N=CC2=CC=C(C=C2)OCC | |
| 281.399 | |
| 281.40 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1218
TSCA : Yes
Recommended Storage : Refrigerator