missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzyl Alcohol 97.0+%, TCI America™

Catalog No. E049225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E049225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E049225G Supplier TCI America Supplier No. E049225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6214-44-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004655
InChI Key UKFLLQIRBABMKF-UHFFFAOYSA-N
Synonym 4-ethoxybenzyl alcohol, 4-ethoxyphenyl methanol, p-ethoxybenzyl alcohol, 4-ethoxybenzylalcohol, benzenemethanol, 4-ethoxy, 4-ethoxy-benzyl alcohol, benzenemethanol,4-ethoxy, 4-ethoxyphenyl methanol #, 4-ethoxyphenyl methan-1-ol, 4-ethyloxy phenyl methanol
PubChem CID 80345
IUPAC Name (4-ethoxyphenyl)methanol
SMILES CCOC1=CC=C(CO)C=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Ethoxybenzyl Alcohol