missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzyl alcohol, 98%

Catalog No. p-7080693
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1033109 10 g
AAA1033118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1033109 Supplier Thermo Scientific Chemicals Supplier No. A1033109

CAS: 6214-44-4 | C9H12O2 | 152.19 g/mol

It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the identification of sulfation sites of metabolites and prediction of the compounds’ biological effects and in chemical investigation of Wedelia calendulacea.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 6214-44-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00004655
InChI Key UKFLLQIRBABMKF-UHFFFAOYSA-N
Synonym 4-ethoxybenzyl alcohol, 4-ethoxyphenyl methanol, p-ethoxybenzyl alcohol, 4-ethoxybenzylalcohol, benzenemethanol, 4-ethoxy, 4-ethoxy-benzyl alcohol, benzenemethanol,4-ethoxy, 4-ethoxyphenyl methanol #, 4-ethoxyphenyl methan-1-ol, 4-ethyloxy phenyl methanol
PubChem CID 80345
IUPAC Name (4-ethoxyphenyl)methanol
SMILES CCOC1=CC=C(CO)C=C1

Specifications

Melting Point 32°C to 34°C
Boiling Point 272°C to 274°C
Flash Point >110°C (230°F)
Refractive Index 1.532
Quantity 10 g
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-Ethoxybenzyl alcohol
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 228-283-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only