missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzeneboronic acid, 98%

CAS: 22237-13-4 | C8H11BO3 | 165.983 g/mol

$52.96 - $524.30

Chemical Identifiers

CAS 22237-13-4
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD00674028
InChI Key WRQNDLDUNQMTCL-UHFFFAOYSA-N
Synonym 4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
PubChem CID 2734351
IUPAC Name (4-ethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC)(O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2368303
View Documents
Thermo Scientific Chemicals
B2368303
1 g
Each for $52.96
Only null left
 
AAB2368306
View Documents
Thermo Scientific Chemicals
B2368306
5 g
Each for $138.05
Only null left
 
AAB2368314
View Documents
Thermo Scientific Chemicals
B2368314
25 g
Each for $524.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22237-13-4
165.983
WRQNDLDUNQMTCL-UHFFFAOYSA-N
2734351
B(C1=CC=C(C=C1)OCC)(O)O
C8H11BO3
MFCD00674028
4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
(4-ethoxyphenyl)boronic acid

Specifications

22237-13-4
C8H11BO3
1 g
4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
B(C1=CC=C(C=C1)OCC)(O)O
165.983
165.99
4-Ethoxybenzeneboronic acid
165°C to 168°C
MFCD00674028
3530408
WRQNDLDUNQMTCL-UHFFFAOYSA-N
(4-ethoxyphenyl)boronic acid
2734351
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only