missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. E004225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004225G 25 g
E0042500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E004225G Supplier TCI America Supplier No. E004225G

Chemical Identifiers

CAS 10031-82-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00003388
InChI Key JRHHJNMASOIRDS-UHFFFAOYSA-N
Synonym benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
PubChem CID 24834
IUPAC Name 4-ethoxybenzaldehyde
SMILES CCOC1=CC=C(C=O)C=C1

Specifications

Color Yellow
Boiling Point 255°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethoxybenzaldehyde
EINECSNumber (9)-0246
RTECSNumber CU6100000
TSCA Yes