accessibility menu, dialog, popup

UserName

CAS RN 10031-82-0

IUPAC Name: 4-ethoxybenzaldehyde
Synonyms: ethoxybenzaldehyde
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: JRHHJNMASOIRDS-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=O)C=C1
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (4)