missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzaldehyde, 99%

Catalog No. p-126146
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174250250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC174250250 Supplier Thermo Scientific Chemicals Supplier No. 174250250
Only null left

CAS: 10031-82-0 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10031-82-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00003388
InChI Key JRHHJNMASOIRDS-UHFFFAOYSA-N
Synonym benzaldehyde, 4-ethoxy, p-ethoxybenzaldehyde, ethoxybenzaldehyde, benzaldehyde, p-ethoxy, homoanisaldehyde, 4-ethyloxy benzaldehyde, unii-gow1h0f49a, fema no. 2413, 4-ethoxy-benzaldehyde, ethoxybenzaldehyde, p
PubChem CID 24834
IUPAC Name 4-ethoxybenzaldehyde
SMILES CCOC1=CC=C(C=O)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 13°C to 14°C
Color Yellow to Brown
Density 1.0800g/mL
Boiling Point 255°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5574 to 1.5594
Linear Formula C2H5OC6H4CHO
Quantity 25 g
Beilstein 08,73
Specific Gravity 1.08
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Ethoxybenzaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-093-3
RTECSNumber CU6100000
TSCA TSCA
Recommended Storage this product darkens on storage

RUO – Research Use Only