missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-methoxybenzaldehyde 98.0+%, TCI America™

Numéro de catalogue. E040625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E040625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. E040625G Fournisseur TCI America Code fournisseur E040625G
Il en reste null

Identifiants chimiques

CAS 120-25-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00016614
InChI Key BERFDQAMXIBOHM-UHFFFAOYSA-N
Synonym 4-ethoxy-3-anisaldehyde, 4-ethoxy-m-anisaldehyde, benzaldehyde, 4-ethoxy-3-methoxy, ethyl-vanillin, asischem n42243, 4-ethoxy-3-methoxy-benzaldehyde, ethylvanillin?, 4-ethoxyvanillin, vanillin ethyl ether, pubchem2648
PubChem CID 67116
IUPAC Name 4-ethoxy-3-methoxybenzaldehyde
SMILES CCOC1=C(C=C(C=C1)C=O)OC

Spécifications

Melting Point 61°C
Color White-Yellow
Boiling Point 167°C
Quantity 25 g
Formula Weight 180.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxy-3-methoxybenzaldehyde