missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2-butanone 92.0+%, TCI America™

Catalog No. E02985ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E02985ML 5 mL
E029825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E02985ML Supplier TCI America Supplier No. E02985ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60044-74-8
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00059372
InChI Key YFBGSHHKHHCVDI-UHFFFAOYSA-N
PubChem CID 536145
IUPAC Name 4-ethoxybutan-2-one
SMILES CCOCCC(=O)C

Specifications

Color Yellow
Boiling Point 153°C
Quantity 5 mL
UN Number 1993
Formula Weight 116.16
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Ethoxy-2-butanone