accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 60044-74-8

IUPAC Name: 4-ethoxybutan-2-one
Synonyms: 4-ethoxybutan-2-one
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: YFBGSHHKHHCVDI-UHFFFAOYSA-N
SMILES: CCOCCC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results