missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Epianhydrotetracycline hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 4465-65-0 |
| Linear Formula | C22H22N2O7 · HCl |
| MDL Number | MFCD00151455 |
| Synonym | 4-Epianhydrotetracycline hydrochloride; (4 R,4 AS,12 AS)-4-(Dimethylamino)-1,4,4 A,5,12,12 A-hexahydro-3,10,11,12 A-tetrahydroxy-6-methyl-1,12-dioxo-2-naphthacenecarboxamide monohydrochloride |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 462.88 |
| Grade | Certified Reference Material |