missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylphosphino)benzoic Acid 97.0+%, TCI America™

Catalog No. D32431G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32431G 1 g
D32435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32431G Supplier TCI America Supplier No. D32431G

Chemical Identifiers

CAS 2129-31-9
Molecular Formula C19H15O2P
Molecular Weight (g/mol) 306.30
MDL Number MFCD00407264
InChI Key GXMHDTPYKRTARV-UHFFFAOYSA-N
Synonym 4-diphenylphosphino benzoic acid, benzoic acid, 4-diphenylphosphino, 4-diphenylphosphanyl-benzoic acid, diphenyl p-carboxyphenyl phosphine, 4-diphenylphosphanyl benzoic acid, 4-diphenylphosphanyl-benzoesaeure, acmc-209fit, 4-diphenylphosphino benzoicacid, p-diphenylphosphino benzoic acid, 4-carboxyphenyl diphenylphosphine
PubChem CID 627824
IUPAC Name 4-(diphenylphosphanyl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 1 g
Formula Weight 306.30
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Diphenylphosphino)benzoic Acid