missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzylamine Dihydrochloride 98.0+%, TCI America™

Catalog No. D16205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16205G 5 g
D162025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16205G Supplier TCI America Supplier No. D16205G
Only null left

Chemical Identifiers

CAS 34403-52-6
Molecular Formula C9H15N2
Molecular Weight (g/mol) 151.23
MDL Number MFCD00012862
InChI Key PDJZOFLRRJQYBF-UHFFFAOYSA-O
PubChem CID 2734633
IUPAC Name [4-(dimethylamino)phenyl]methanaminium
SMILES CN(C)C1=CC=C(C[NH3+])C=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 223.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzylamine Dihydrochloride