missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzoic anhydride, 97%

Catalog No. p-7031430
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3250303 1 g
AAH3250306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3250303 Supplier Thermo Scientific Chemicals Supplier No. H3250303

CAS: 7474-31-9 | C18H20N2O3 | 312.369 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7474-31-9
Molecular Formula C18H20N2O3
Molecular Weight (g/mol) 312.369
MDL Number MFCD09757554
InChI Key KGUATYWLOBCNMI-UHFFFAOYSA-N
Synonym 4-dimethylaminobenzoic anhydride, 4-dimethylamino benzoyl 4-dimethylamino benzoate, acmc-20amjw, benzoic acid, 4-dimethylamino-, anhydride, 4-dimethylamino phenyl anhydride #, bis 4-dimethylamino benzoic anhydride, benzoic acid,4-dimethylamino-, anhydride with 4-dimethylamino benzoic acid
PubChem CID 344732
IUPAC Name [4-(dimethylamino)benzoyl] 4-(dimethylamino)benzoate
SMILES CN(C)C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)N(C)C

Specifications

Melting Point 149°C to 154°C
Quantity 1 g
Formula Weight 312.37
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminobenzoic anhydride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only