accessibility menu, dialog, popup

UserName

CH3NH3COOONCH3H3C

CAS RN 7474-31-9

IUPAC Name: 4-(dimethylamino)benzoyl 4-(dimethylamino)benzoate
Synonyms: 4-(dimethylamino)benzoyl 4-(dimethylamino)benzoate
Molecular Weight (g/mol): 312.37
Molecular Formula: C18H20N2O3
InChi Key: KGUATYWLOBCNMI-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C=C1)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results