missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzenediazosulfonic Acid Sodium Salt 97.0+%, TCI America™

Catalog No. D13945G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D13945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D13945G Supplier TCI America Supplier No. D13945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent used for research and experimental applications.

Chemical Identifiers

CAS 140-56-7
Molecular Formula C8H10N3NaO3S
Molecular Weight (g/mol) 251.24
MDL Number MFCD00059900
InChI Key IWDQPCIQCXRBQP-UHFFFAOYSA-M
Synonym fenaminosulf, dexon, phenaminosulf, deksonal, dexoxon, lesan, formulated fenaminosulf, gold orange mp, sodium fenaminosulf, dexon pesticide
PubChem CID 23668198
ChEBI CHEBI:82078
IUPAC Name sodium N-{[4-(dimethylamino)phenyl]imino}sulfamate
SMILES [Na+].CN(C)C1=CC=C(C=C1)N=NS([O-])(=O)=O

Specifications

Quantity 5 g
UN Number 2811
Formula Weight 251.24
Percent Purity ≥97.0% (HPLC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminobenzenediazosulfonic Acid Sodium Salt
RTECSNumber CZ1750000
TSCA No