accessibility menu, dialog, popup

UserName

CAS RN 140-56-7

IUPAC Name: sodium N-{[4-(dimethylamino)phenyl]imino}sulfamate
Synonyms: sodium N-{[4-(dimethylamino)phenyl]imino}sulfamate
Molecular Weight (g/mol): 251.24
Molecular Formula: C8H10N3NaO3S
InChi Key: IWDQPCIQCXRBQP-UHFFFAOYSA-M
SMILES: [Na+].CN(C)C1=CC=C(C=C1)N=NS([O-])(=O)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results