missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

$129.62 - $129.62

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC149281000
SDS View Documents
Thermo Scientific Chemicals
149281000
100 g
Each for $129.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-10-7
149.19
BGNGWHSBYQYVRX-UHFFFAOYSA-N
7479
CN(C)C1=CC=C(C=O)C=C1
C9H11NO
MFCD00003381
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
4-(dimethylamino)benzaldehyde

Specifications

100-10-7
100.0
White to Off-white
Authentic
C9H11NO
MFCD00003381
14, 31
15, 3252
BGNGWHSBYQYVRX-UHFFFAOYSA-N
4-(dimethylamino)benzaldehyde
7479
≥97.5%
4-Dimethylaminobenzaldehyde, 98%
97.5
70°C to 75°C
5.15
98%
(CH3)2NC6H4CHO
100 g
01,273; 02,146
4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
CN(C)C1=CC=C(C=O)C=C1
149.19
149.19
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.

Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . Harmful to aquatic organisms, may cause long-term adverse effects in the aquatic environment.

Signal Word: WARNING!

EINECSNumber : 202-819-0

RTECSNumber : CU5775000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only