missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)pyridine N-Oxide Hydrate 98.0+%, TCI America™

Catalog No. d32205g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32205G 5 g
D32201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32205G Supplier TCI America Supplier No. D32205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1005-31-8
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
InChI Key WZMNQOYCHMGCSS-UHFFFAOYSA-N
Synonym DMAPO
PubChem CID 136795
IUPAC Name N,N-dimethyl-1-oxidopyridin-1-ium-4-amine
SMILES CN(C)C1=CC=[N+](C=C1)[O-]

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)pyridine N-Oxide Hydrate