accessibility menu, dialog, popup

UserName

CAS RN 1005-31-8

IUPAC Name: 4-(dimethylamino)pyridin-1-ium-1-olate
Synonyms: 4-(dimethylamino)pyridin-1-ium-1-olate
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: WZMNQOYCHMGCSS-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=[N+]([O-])C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results