missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$219.95 - $219.95
Specifications
| CAS | 28611-39-4 |
|---|---|
| Melting Point | 227°C |
| Color | White-Yellow |
| Molecular Formula | C8H12BNO2 |
| MDL Number | MFCD01074642 |
Specifications
| 28611-39-4 | |
| White-Yellow | |
| MFCD01074642 | |
| [4-(dimethylamino)phenyl]boronic acid | |
| 165.00 |
| 227°C | |
| C8H12BNO2 | |
| RIIPFHVHLXPMHQ-UHFFFAOYSA-N | |
| 2734344 | |
| Crystalline Powder |