missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)benzyl Alcohol 98.0+%, TCI America™
$521.94 - $1776.44
Chemical Identifiers
| CAS | 1703-46-4 |
|---|---|
| Molecular Formula | C9H13NO |
| Molecular Weight (g/mol) | 151.21 |
| MDL Number | MFCD03840449 |
| InChI Key | WQBCAASPALGAKX-UHFFFAOYSA-N |
| Synonym | 4-Hydroxymethyl-N,N-dimethylaniline |
| PubChem CID | 137166 |
| IUPAC Name | [4-(dimethylamino)phenyl]methanol |
| SMILES | CN(C)C1=CC=C(CO)C=C1 |
Chemical Identifiers
| 1703-46-4 | |
| 151.21 | |
| WQBCAASPALGAKX-UHFFFAOYSA-N | |
| 137166 | |
| CN(C)C1=CC=C(CO)C=C1 |
| C9H13NO | |
| MFCD03840449 | |
| 4-Hydroxymethyl-N,N-dimethylaniline | |
| [4-(dimethylamino)phenyl]methanol |
Specifications
| 1703-46-4 | |
| 123°C | |
| MFCD03840449 | |
| 4-Hydroxymethyl-N,N-dimethylaniline | |
| CN(C)C1=CC=C(CO)C=C1 | |
| 151.21 | |
| 151.21 | |
| Liquid |
| Yellow | |
| C9H13NO | |
| 5 g | |
| WQBCAASPALGAKX-UHFFFAOYSA-N | |
| [4-(dimethylamino)phenyl]methanol | |
| 137166 | |
| ≥98.0% (T) | |
| 4-(Dimethylamino)benzyl Alcohol |
Safety and Handling
TSCA : No
Recommended Storage : Freezer