missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzaldehyde, ≥99% (HPLC), Solstice

Catalog No. p-7120091
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046742 50 g Glass Bottle
60046741 250 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046742 Supplier Solstice Supplier No. 3907050G

for the determination of hydroxyproline,≥99.0% (HPLC)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonal
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 73°C to 75°C
Color White
Boiling Point 176°C to 177°C
Flash Point 164°C (327.2°F)
Ignition Residue ≤0.1% (as SO4)
Loss on Drying 0 to 0.5%
Packaging Glass Bottle
Linear Formula (CH3)2NC6H4CHO
Quantity 50 g
UN Number NONH for all modes of transport
Beilstein 606802
Formula Weight 149.19g/mol
Percent Purity ≥99% (HPLC)
Chemical Name or Material 4-(Dimethylamino)benzaldehyde
Show More Show Less
RTECSNumber 202-819-0