missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethylamino-1-neopentylpyridinium Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 228.76 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Dimethylamino-1-neopentylpyridinium Chloride |
Safety and Handling
| TSCA | Yes |