accessibility menu, dialog, popup

UserName

H3CH3CCH3NNCH3H3CCl

CAS RN 109911-77-5

IUPAC Name: 4-(dimethylamino)-1-(2,2-dimethylpropyl)pyridin-1-ium chloride
Synonyms: 4-(dimethylamino)-1-(2,2-dimethylpropyl)pyridin-1-ium chloride
Molecular Weight (g/mol): 228.76
Molecular Formula: C12H21ClN2
InChi Key: OTCQGJASYOVVCB-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=[N+](CC(C)(C)C)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results