missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-1-naphthyl Isothiocyanate 98.0+%, TCI America™

Catalog No. D16691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16691G 1 g
D16695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16691G Supplier TCI America Supplier No. D16691G
Only null left

Chemical Identifiers

CAS 29711-79-3
Molecular Formula C13H12N2S
Molecular Weight (g/mol) 228.31
MDL Number MFCD00003883
InChI Key SMZHGTXTWIAKGS-UHFFFAOYSA-N
Synonym 4-dimethylamino-1-naphthylisothiocyanate, 4-dimethylamino-1-naphthyl isothiocyanate, danito, 4-n,n-dimethylamino-1-naphthylisothiocyanate, n-4-isothiocyanato-1-naphthyl-n,n-dimethylamine, 4-isothiocyanato-n,n-dimethyl-1-naphthalenamine, 4-dimethylamino naphthalenisothiocyanate, acmc-209umu, 4-dimethylamino-1-naphthyl 1sothiocyanate
PubChem CID 122465
IUPAC Name 4-isothiocyanato-N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=C2C=CC=CC2=C(C=C1)N=C=S

Specifications

Melting Point 67°C
Color Yellow
Quantity 1 g
Formula Weight 228.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylamino-1-naphthyl Isothiocyanate [for HPLC Labeling]