missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino-1-naphthoic acid, 98%

Catalog No. p-7044721
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0794403 1 g
AAL0794406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0794403 Supplier Thermo Scientific Chemicals Supplier No. L0794403

CAS: 78062-03-0 | C13H13NO2 | 215.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78062-03-0
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.252
MDL Number MFCD00051797
InChI Key LZQKNRRVTOJUHQ-UHFFFAOYSA-N
Synonym 4-dimethylamino-1-naphthoic acid, 4-dimethylamino naphthalene-1-carboxylic acid, 4-dimethylamino-1-naphthalenecarboxylic acid, 4-dimethylamino naphthalic acid, 4-dimethylamino-1-naphthoicacid, 4-dimethylamino-1-naphthoic acid #, 4-dimethylaminonaphthalenecarboxylic acid, 4-dimethylamino naphthalenecarboxylic acid, 4-dimethylamino-naphthalene-1-carboxylic acid
PubChem CID 522695
IUPAC Name 4-(dimethylamino)naphthalene-1-carboxylic acid
SMILES CN(C)C1=CC=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point ∽160°C (decomposition)
Quantity 1 g
Beilstein 2723206
Formula Weight 215.25
Percent Purity 98%
Chemical Name or Material 4-Dimethylamino-1-naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only