missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Difluoromethoxy)toluene 96.0+%, TCI America™
$106.25 - $106.25
Chemical Identifiers
| CAS | 1583-83-1 |
|---|---|
| Molecular Formula | C8H8F2O |
| Molecular Weight (g/mol) | 158.148 |
| MDL Number | MFCD00042253 |
| InChI Key | DJDQNISEJVPQCS-UHFFFAOYSA-N |
| Synonym | alpha,alpha-Difluoro-4-methylanisole |
| PubChem CID | 74104 |
| IUPAC Name | 1-(difluoromethoxy)-4-methylbenzene |
| SMILES | CC1=CC=C(C=C1)OC(F)F |
Chemical Identifiers
| 1583-83-1 | |
| 158.148 | |
| DJDQNISEJVPQCS-UHFFFAOYSA-N | |
| 74104 | |
| CC1=CC=C(C=C1)OC(F)F |
| C8H8F2O | |
| MFCD00042253 | |
| alpha,alpha-Difluoro-4-methylanisole | |
| 1-(difluoromethoxy)-4-methylbenzene |
Specifications
| 1583-83-1 | |
| 167°C | |
| MFCD00042253 | |
| alpha,alpha-Difluoro-4-methylanisole | |
| CC1=CC=C(C=C1)OC(F)F | |
| 158.148 | |
| 158.15 | |
| Liquid |
| Yellow | |
| C8H8F2O | |
| 1 g | |
| DJDQNISEJVPQCS-UHFFFAOYSA-N | |
| 1-(difluoromethoxy)-4-methylbenzene | |
| 74104 | |
| ≥96.0% (GC) | |
| 4-(Difluoromethoxy)toluene |
Safety and Handling
TSCA : No