missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)benzonitrile 98.0+%, TCI America™

Catalog No. D49555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49555G 5 g
D49551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49555G Supplier TCI America Supplier No. D49555G
Only null left

Chemical Identifiers

CAS 90446-25-6
Molecular Formula C8H5F2NO
Molecular Weight (g/mol) 169.131
MDL Number MFCD00085006
InChI Key OGMOYCCCAFJQKI-UHFFFAOYSA-N
Synonym 1-Cyano-4-(difluoromethoxy)benzene
PubChem CID 2736990
IUPAC Name 4-(difluoromethoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC(F)F

Specifications

Melting Point 42°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 169.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Difluoromethoxy)benzonitrile